Methyl-3-(aminomethyl)benzoate hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 3-(aminomethyl)benzoate hydrochloride

OR7914-1G OR7914-5G OR7914-25G
APOSOR7914-1GEA 29 EUR
APOSOR7914-1G APOSOR7914-5G APOSOR7914-25G
Methyl-3-(aminomethyl)benzoate hydrochloride
Methyl-3-(aminomethyl)benzoate hydrochloride
Formel: C₉H₁₂ClNO₂
Schmelzpunkt: 179…181 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04039173
CAS-Nummer: 17841-68-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR