1,2-Dimethyl-1H-indole-3-carbonitrile 95+%

Lieferant: Apollo Scientific
Achtung

OR7926-250MG
APOSOR7926-250MGEA 21.9 EUR
APOSOR7926-250MG
1,2-Dimethyl-1H-indole-3-carbonitrile 95+%
1,2-Dimethyl-1H-indole-3-carbonitrile
Formel: C₁₁H₁₀N₂
Schmelzpunkt: 103…105 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00460785
CAS-Nummer: 51072-84-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR