6-(Methylsulphonyl)-1-(oxiran-2-ylmethoxy)-1H-indole 95+%

Lieferant: Apollo Scientific

OR7927-1G
APOSOR7927-1GEA 185 EUR
APOSOR7927-1G
6-(Methylsulphonyl)-1-(oxiran-2-ylmethoxy)-1H-indole 95+%
6-(Methylsulphonyl)-1-(oxiran-2-ylmethoxy)-1H-indole
Formel: C₁₂H₁₃NO₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD00203135
CAS-Nummer: 849924-91-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR