6-[(Piperazin-1-yl)carbonyl]-1H-indole 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: (1H-Indol-6-yl)(piperazin-1-yl)methanone

OR7928-250MG
APOSOR7928-250MGEA 65.5 EUR
APOSOR7928-250MG
6-[(Piperazin-1-yl)carbonyl]-1H-indole 95+%
6-[(Piperazin-1-yl)carbonyl]-1H-indole
Formel: C₁₃H₁₅N₃O
Molecular Weight: 229,28 g/mol
Schmelzpunkt: 173…176 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01765613
CAS-Nummer: 633322-11-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR