1,3-Dihydro-1-(piperidin-4-yl)-(2H)-indol-2-one

Lieferant: Apollo Scientific
Achtung

OR7929-1G
APOSOR7929-1GEA 154 EUR
APOSOR7929-1G
1,3-Dihydro-1-(piperidin-4-yl)-(2H)-indol-2-one
1,3-Dihydro-1-(piperidin-4-yl)-(2H)-indol-2-one
Formel: C₁₃H₁₆N₂O
Molecular Weight: 216,29 g/mol
Schmelzpunkt: 163…166 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03426085
CAS-Nummer: 16223-25-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR