2-Bromo-5-methylsulphonylaniline
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Bromo-5-(methylsulphonyl)aniline
OR7945-1G
APOSOR7945-1GEA
164
EUR
APOSOR7945-1G
2-Bromo-5-methylsulphonylaniline
2-Bromo-5-methylsulphonylaniline
Formel:
C₇H₈BrNO₂S Molecular Weight: 250,12 g/mol Schmelzpunkt: 117…118 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD09027715 CAS-Nummer: 942474-24-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...