1-Phenyl-2-pyrazin-2-ylethanone 95+%

Lieferant: Apollo Scientific
Achtung

OR7947-1G
APOSOR7947-1GEA 190 EUR
APOSOR7947-1G
1-Phenyl-2-pyrazin-2-ylethanone 95+%
1-Phenyl-2-pyrazin-2-ylethanone
Formel: C₁₂H₁₀N₂O
Molecular Weight: 198,23 g/mol
Siedepunkt: 80…82 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04037906
CAS-Nummer: 40061-45-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR