6-Chloro-2-methylindole 98%
Lieferant: Apollo Scientific
Synonyme:
6-Chloro-2-methyl-1H-indole
OR7950-250MG
APOSOR7950-250MGEA
63.8
EUR
APOSOR7950-250MG
6-Chloro-2-methylindole 98%
6-Chloro-2-methylindole
Formel:
C₉H₈ClN Molecular Weight: 165.62 g/mol Schmelzpunkt: 75…77 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09027716 CAS-Nummer: 6127-17-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...