1-(Bromomethyl)-2-chloro-4-(methylsulphonyl)benzene 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chloro-4-(methylsulphonyl)benzyl bromide

OR7956-100MG
APOSOR7956-100MGEA 21.9 EUR
APOSOR7956-100MG
1-(Bromomethyl)-2-chloro-4-(methylsulphonyl)benzene 95+%
1-(Bromomethyl)-2-chloro-4-(methylsulphonyl)benzene
Formel: C₈H₈BrClO₂S
Molecular Weight: 283,58 g/mol
Schmelzpunkt: 119…121 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04037959
CAS-Nummer: 180200-86-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR