1-(Bromomethyl)-4-chloro-2-(methylsulphonyl)benzene

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Chloro-2-(methylsulphonyl)benzyl bromide

OR7957-100MG
APOSOR7957-100MGEA 21.9 EUR
APOSOR7957-100MG
1-(Bromomethyl)-4-chloro-2-(methylsulphonyl)benzene
1-(Bromomethyl)-4-chloro-2-(methylsulphonyl)benzene
Formel: C₈H₈BrClO₂S
Molecular Weight: 283,58 g/mol
Schmelzpunkt: 113…115 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04037923
CAS-Nummer: 849035-64-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR