1H-Indole-2-carbohydrazide

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Hydrazinocarbonyl)-1H-indole, 1H-Indole-2-carboxylic acid hydrazide

OR7963-1G
APOSOR7963-1GEA 97.3 EUR
APOSOR7963-1G
1H-Indole-2-carbohydrazide
1H-Indole-2-carbohydrazide
Formel: C₉H₉N₃O
Molecular Weight: 175,187 g/mol
Schmelzpunkt: 243…245 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02293210
CAS-Nummer: 5055-39-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR