3-Nitro-o-toluylsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Methyl-3-nitrobenzoesäure
OR7971-500G
APOSOR7971-500GEA
60.9
EUR
APOSOR7971-500G
3-Nitro-o-toluylsäure
3-Nitro-o-toluylsäure
Formel:
C₈H₇NO₄ Molecular Weight: 181.15 g/mol Schmelzpunkt: 182…186 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007160 CAS-Nummer: 1975-50-4 EINECS: 217-826-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...