1,2-Dibromo-4-(methylsulphonyl)benzene 95+%

Lieferant: Apollo Scientific

OR7977-1G OR7977-5G OR7977-25G
APOSOR7977-1GEA 128 EUR
APOSOR7977-1G APOSOR7977-5G APOSOR7977-25G
1,2-Dibromo-4-(methylsulphonyl)benzene 95+%
1,2-Dibromo-4-(methylsulphonyl)benzene
Formel: C₇H₆Br₂O₂S
Schmelzpunkt: 141…142 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04037933
CAS-Nummer: 849035-70-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR