1,2-Dichloro-4-methyl-5-(methylsulphonyl)-3-nitrobenzene
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4,5-Dichloro-2-methyl-3-nitrophenyl methyl sulphone
OR7981-1G
APOSOR7981-1GEA
232
EUR
APOSOR7981-1G
1,2-Dichloro-4-methyl-5-(methylsulphonyl)-3-nitrobenzene
1,2-Dichloro-4-methyl-5-(methylsulphonyl)-3-nitrobenzene
Formel:
C₈H₇Cl₂NO₄S Schmelzpunkt: 163…165 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04037949 CAS-Nummer: 849035-79-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...