3-Methylsulphonylbenzenesulphonyl chloride 95%
Lieferant: Apollo Scientific
|
Gefahr
|
OR7993-1G
OR7993-25G
OR7993-5G
APOSOR7993-1GEA
0
EUR
APOSOR7993-1G
APOSOR7993-25G
APOSOR7993-5G
3-Methylsulphonylbenzenesulphonyl chloride 95%
3-Methylsulphonylbenzenesulphonyl chloride
Formel:
C₇H₇ClO₄S₂ Schmelzpunkt: 93…96 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04037968 CAS-Nummer: 5335-40-0 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...