Cäsiumchlorid 99.99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Cesium Chloride
OR8001T-100G
OR8001T-1KG
OR8001T-25G
APOSOR8001T-100GEA
234
EUR
APOSOR8001T-100G
APOSOR8001T-1KG
APOSOR8001T-25G
Cäsiumchlorid 99.99%
Cesium chloride
For molecular genetic applications. Used in preparingnucleic acids for subsequent hybridization or cloning experiments.
Formel:
CsCl Molecular Weight: 168.36 g/mol Siedepunkt: 1303 Schmelzpunkt: 646 °C Dichte: 3.988 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010955 CAS-Nummer: 7647-17-8 EINECS: 231-600-2 Merck Index: 13,02021 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...