Cäsiumchlorid 99.99%

Lieferant: Apollo Scientific
Achtung

OR8001T-100G OR8001T-1KG OR8001T-25G
APOSOR8001T-100GEA 234 EUR
APOSOR8001T-100G APOSOR8001T-1KG APOSOR8001T-25G
Cäsiumchlorid 99.99%
Cesium chloride

For molecular genetic applications. Used in preparingnucleic acids for subsequent hybridization or cloning experiments.

Formel: CsCl
Schmelzpunkt: 646 °C
Dichte: 3,988 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00010955
CAS-Nummer: 7647-17-8
EINECS: 231-600-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR