Ethyl 4,6-dichloro-2-(methylthio)quinoline-3-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 4,6-dichloro-2-(methylsulphanyl)quinoline, 4,6-Dichloro-3-(ethoxycarbonyl)-2-(methylthio)quinoline

OR8002-1G OR8002-500MG
APOSOR8002-1GEA 105 EUR
APOSOR8002-1G APOSOR8002-500MG
Ethyl 4,6-dichloro-2-(methylthio)quinoline-3-carboxylate
Ethyl 4,6-dichloro-2-(methylthio)quinoline-3-carboxylate
Formel: C₁₃H₁₁Cl₂NO₂S
Schmelzpunkt: 133…135 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00276616
CAS-Nummer: 227958-96-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR