(S)-3-((tert-Butoxycarbonyl)amino)-2-(4-chlorophenyl)propanoic acid 95%

Lieferant: Apollo Scientific
Achtung

OR80059 OR80059-100MG OR80059-1G
APOSOR80059EA 478 EUR
APOSOR80059 APOSOR80059-100MG APOSOR80059-1G
(S)-3-((tert-Butoxycarbonyl)amino)-2-(4-chlorophenyl)propanoic acid 95%
(S)-3-((tert-Butoxycarbonyl)amino)-2-(4-chlorophenyl)propanoic acid
Formel: C₁₄H₁₈ClNO₄
MDL: MFCD16394572
CAS-Nummer: 1001180-04-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR