2-(2-Amino-4-methylthien-3-yl)-1,3-benzothiazole 97%

Lieferant: Apollo Scientific
Achtung

OR8011-100MG
APOSOR8011-100MGEA 33.5 EUR
APOSOR8011-100MG
2-(2-Amino-4-methylthien-3-yl)-1,3-benzothiazole 97%
2-(2-Amino-4-methylthien-3-yl)-1,3-benzothiazole
Formel: C₁₂H₁₀N₂S₂
Molecular Weight: 246,351 g/mol
Schmelzpunkt: 110…112 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02179980
CAS-Nummer: 232941-00-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR