2-(1,2-Benzisoxazol-3-yl)essigsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1,2-Benzisoxazole-3-acetic Acid

OR8018-100G OR8018-1G
APOSOR8018-100GEA 180 EUR
APOSOR8018-100G APOSOR8018-1G
2-(1,2-Benzisoxazol-3-yl)essigsäure 97%
2-(1,2-Benzisoxazol-3-yl)essigsäure
Formel: C₉H₇NO₃
Molecular Weight: 177.1568 g/mol
Schmelzpunkt: 123…129 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02180388
CAS-Nummer: 4865-84-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR