4-(Benzyloxy)anilin Hydrochlorid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Benzyloxyanilinium Chloride, 4-Aminophenyl benzyl ether hydrochloride, p-Benzyloxyaniline hydrochloride
OR8024-25G
APOSOR8024-25GEA
21.9
EUR
APOSOR8024-25G
4-(Benzyloxy)anilin Hydrochlorid
4-(Benzyloxy)anilin Hydrochlorid
Formel:
C₁₃H₁₄ClNO Molecular Weight: 235,711 g/mol Schmelzpunkt: 227…229 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012995 CAS-Nummer: 51388-20-6 EINECS: 257-170-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...