Boc-Thz-OH 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Boc-L-thiazolidine-4-carboxylic acid

OR8031-100G
APOSOR8031-100GEA 97.3 EUR
APOSOR8031-100G
Boc-Thz-OH 98%
Boc-Thz-OH
Formel: C₉H₁₅NO₄S
Schmelzpunkt: 132…136 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00077002
CAS-Nummer: 51077-16-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR