1-(1,3-Benzodioxol-5-yl)-2-bromo-1-ethanone
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
5-(Bromoacetyl)-1,3-benzodioxole
OR8032-250MG
APOSOR8032-250MGEA
40.6
EUR
APOSOR8032-250MG
1-(1,3-Benzodioxol-5-yl)-2-bromo-1-ethanone
1-(1,3-Benzodioxol-5-yl)-2-bromo-1-ethanone
Formel:
C₉H₇BrO₃ Molecular Weight: 243.06 g/mol Schmelzpunkt: 91…94 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01910077 CAS-Nummer: 40288-65-1 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...