3-(1-Methyl-1H-pyrazol-3-yl)aniline
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(3-Aminophenyl)-1-methyl-1H-pyrazole
OR8034-1G
APOSOR8034-1GEA
190
EUR
APOSOR8034-1G
3-(1-Methyl-1H-pyrazol-3-yl)aniline
3-(1-Methyl-1H-pyrazol-3-yl)aniline
Formel:
C₁₀H₁₁N₃ Molecular Weight: 173,214 g/mol Schmelzpunkt: 116…119 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00084885 CAS-Nummer: 175202-37-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...