4-Bromo-1-(4-chlorobenzyl)-1H-pyrazole-5-carboxaldehyde
Lieferant: Apollo Scientific
OR8036-1G
APOSOR8036-1GEA
437
EUR
APOSOR8036-1G
4-Bromo-1-(4-chlorobenzyl)-1H-pyrazole-5-carboxaldehyde
4-Bromo-1-(4-chlorobenzyl)-1H-pyrazole-5-carboxaldehyde
Formel:
C₁₁H₈BrClN₂O Molecular Weight: 299,551 g/mol Schmelzpunkt: 115…118 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00103038 CAS-Nummer: 289504-53-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...