tert-Butyl-4-carbamothioylpiperidine-1-carboxylate 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR8046-1G
OR8046-5G
APOSOR8046-1GEA
21.9
EUR
APOSOR8046-1G
APOSOR8046-5G
tert-Butyl-4-carbamothioylpiperidine-1-carboxylate 95%
tert-Butyl-4-carbamothioylpiperidine-1-carboxylate
Formel:
C₁₁H₂₀N₂O₂S Schmelzpunkt: 122…124 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02180954 CAS-Nummer: 214834-18-1 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...