4,5-Dichloro-6-methyl-2-(pyridin-2-yl)pyrimidine

Lieferant: Apollo Scientific

OR8074-1G
APOSOR8074-1GEA 125 EUR
APOSOR8074-1G
4,5-Dichloro-6-methyl-2-(pyridin-2-yl)pyrimidine
4,5-Dichloro-6-methyl-2-(pyridin-2-yl)pyrimidine
Formel: C₁₀H₇Cl₂N₃
Molecular Weight: 240,089 g/mol
Schmelzpunkt: 132…133 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00097945
CAS-Nummer: 306935-55-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR