Fmoc-d-pipecolic acid 97%

Lieferant: Apollo Scientific

OR8101-5G
APOSOR8101-5GEA 0 EUR
APOSOR8101-5G
Fmoc-d-pipecolic acid 97%
Fmoc-d-pipecolic acid
Formel: C₂₁H₂₁NO₄
Schmelzpunkt: 149…153 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00235899
CAS-Nummer: 101555-63-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR