(3S)-1,2,3,4-Tetrahydro-3-isochinolinylmethanol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(S)-(-)-1,2,3,4-Tetrahydro-3-isoquinolinemethanol, (S)-(1,2,3,4-Tetrahydroisoquinolin-3-yl)methanol, (S)-3-Hydroxymethyl-1,2,3,4-tetrahydroisoquinoline
OR8104-1G
OR8104-5G
OR8104-25G
APOSOR8104-1GEA
21.9
EUR
APOSOR8104-1G
APOSOR8104-5G
APOSOR8104-25G
(3S)-1,2,3,4-Tetrahydro-3-isochinolinylmethanol 97%
(S)-1,2,3,4-Tetrahydroisoquinoline-3-methanol
Formel:
C₁₀H₁₃NO Schmelzpunkt: 114…116 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631316 CAS-Nummer: 18881-17-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...