6-[(2R,3R,4R,5S)-2-(Hydroxymethyl)-3,4,5-trihydroxypiperidin-1-yl]hexanoic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: N-(5-Carboxypent-1-yl)-1-deoxynojirimycin

OR8120T-5MG
APOSOR8120T-5MGEA 219 EUR
APOSOR8120T-5MG
6-[(2R,3R,4R,5S)-2-(Hydroxymethyl)-3,4,5-trihydroxypiperidin-1-yl]hexanoic acid
6-[(2R,3R,4R,5S)-2-(Hydroxymethyl)-3,4,5-trihydroxypiperidin-1-yl]hexanoic acid

Ligand used in the preparation of an affinity resin highlyspecific for glucosidase I purification. Glucosidase I isinvolved in the processing of N-linked oilgosaccharides.

Formel: C₁₂H₂₃NO₆
Molecular Weight: 277,314 g/mol
Schmelzpunkt: 185…187 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD01320383
CAS-Nummer: 79206-51-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR