Methyl 5-(chlorosulphonyl)-2-methyl-3-furoate 95+%

Lieferant: Apollo Scientific
Gefahr

Synonyme: Methyl 5-(chlorosulphonyl)-2-methylfuran-3-carboxylate, 5-(Chlorosulphonyl)-3-(methoxycarbonyl)-2-methylfuran

OR8121-250MG
APOSOR8121-250MGEA 174 EUR
APOSOR8121-250MG
Methyl 5-(chlorosulphonyl)-2-methyl-3-furoate 95+%
Methyl 5-(chlorosulphonyl)-2-methyl-3-furoate
Formel: C₇H₇ClO₅S
Molecular Weight: 238,645 g/mol
Schmelzpunkt: 35…38 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02180742
CAS-Nummer: 306936-35-6
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR