6-Bromimidazo[1,2-a]pyrimidin

Lieferant: Apollo Scientific
Achtung

OR8122-1G OR8122-250MG OR8122-5G
APOSOR8122-1GEA 31.9 EUR
APOSOR8122-1G APOSOR8122-250MG APOSOR8122-5G
6-Bromimidazo[1,2-a]pyrimidin
6-Bromoimidazo[1,2-a]pyrimidine
Formel: C₆H₄BrN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09261434
CAS-Nummer: 865156-68-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR