Methyl 5-(chlorosulphonyl)-1-methyl-1H-pyrrole-2-carboxylate

Lieferant: Apollo Scientific
Gefahr

Synonyme: 5-(Ethoxycarbonyl)-1-methyl-1H-pyrrole-2-sulphonyl chloride, 2-(Chlorosulphonyl)-5-(methoxycarbonyl)-1-methyl-1H-pyrrole

OR8129-250MG
APOSOR8129-250MGEA 634 EUR
APOSOR8129-250MG
Methyl 5-(chlorosulphonyl)-1-methyl-1H-pyrrole-2-carboxylate
Methyl 5-(chlorosulphonyl)-1-methyl-1H-pyrrole-2-carboxylate
Formel: C₇H₈ClNO₄S
Molecular Weight: 237,661 g/mol
Schmelzpunkt: 79…80 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02180789
CAS-Nummer: 306936-53-8
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR