2-[4-(4-Nitrophenyl)-1,3-thiazol-2-yl]acetonitrile 97%

Lieferant: Apollo Scientific

OR8141-1G
APOSOR8141-1GEA 125 EUR
APOSOR8141-1G
2-[4-(4-Nitrophenyl)-1,3-thiazol-2-yl]acetonitrile 97%
2-[4-(4-Nitrophenyl)-1,3-thiazol-2-yl]acetonitrile
Formel: C₁₁H₇N₃O₂S
Schmelzpunkt: 147…148 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00724226
CAS-Nummer: 69625-13-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR