4-Amino-2-(ethylthio)-5-(hydroxymethyl)pyrimidine 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR8157-1G
APOSOR8157-1GEA
132
EUR
APOSOR8157-1G
4-Amino-2-(ethylthio)-5-(hydroxymethyl)pyrimidine 98%
4-Amino-2-(ethylthio)-5-(hydroxymethyl)pyrimidine
Formel:
C₇H₁₁N₃OS Molecular Weight: 185,247 g/mol Schmelzpunkt: 158…159 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00090796 CAS-Nummer: 98432-26-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...