2-(Benzyloxycarbonylamino)glutarimide 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Benzyl (2,6-dioxopiperidin-3-yl)carbamate

OR8165-1G
APOSOR8165-1GEA 277 EUR
APOSOR8165-1G
2-(Benzyloxycarbonylamino)glutarimide 98%
2-(Benzyloxycarbonylamino)glutarimide
Formel: C₁₃H₁₄N₂O₄
Molecular Weight: 262,261 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01075195
CAS-Nummer: 24666-55-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR