4-(Benzyloxy)-3-methylbutanoic acid 96+%

Lieferant: Apollo Scientific
Achtung

OR8167-1G
APOSOR8167-1GEA 144 EUR
APOSOR8167-1G
4-(Benzyloxy)-3-methylbutanoic acid 96+%
4-(Benzyloxy)-3-methylbutanoic acid
Formel: C₁₂H₁₆O₃
Molecular Weight: 208,254 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01075210
CAS-Nummer: 132437-90-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR