1-Brom-4-phenylbutan

Lieferant: Apollo Scientific
Achtung

OR8178-1G
APOSOR8178-1GEA 21.9 EUR
APOSOR8178-1G
1-Brom-4-phenylbutan
1-Brom-4-phenylbutan
Formel: C₁₀H₁₃Br
Lagertemperatur: Raumtemperatur
MDL: MFCD00154988
CAS-Nummer: 13633-25-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR