(5-Brompentyl)benzol 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Brom-5-phenylpentan
OR8183-5G
APOSOR8183-5GEA
318
EUR
APOSOR8183-5G
(5-Brompentyl)benzol 95+%
(5-Bromopentyl)benzene
Formel:
C₁₁H₁₅Br Molecular Weight: 227.14 g/mol Siedepunkt: 148 °C/18mmHg Dichte: 1.23 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD01075177 CAS-Nummer: 14469-83-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...