2-[3-(Ethoxycarbonyl)piperidin-1-yl]-5-nitrobenzoic acid

Lieferant: Apollo Scientific
Achtung

OR8192-1G
APOSOR8192-1GEA 190 EUR
APOSOR8192-1G
2-[3-(Ethoxycarbonyl)piperidin-1-yl]-5-nitrobenzoic acid
2-[3-(Ethoxycarbonyl)piperidin-1-yl]-5-nitrobenzoic acid
Formel: C₁₅H₁₈N₂O₆
Schmelzpunkt: 127…128 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09064901
CAS-Nummer: 1000018-70-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR