Benzyl-α-D-mannopyranoside

Lieferant: Apollo Scientific

OR8200T-1G
APOSOR8200T-1GEA 132 EUR
APOSOR8200T-1G
Benzyl-α-D-mannopyranoside
Benzyl-α-D-mannopyranoside
Formel: C₁₃H₁₈O₆
Molecular Weight: 270,278 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00153924
CAS-Nummer: 15548-45-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR