δ-Bromobutylhydantoin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-(4-Bromobutyl)-2,4-imidazolidinedione, δ-Bromobutylhydantoin
OR8201T-250MG
APOSOR8201T-250MGEA
0
EUR
APOSOR8201T-250MG
δ-Bromobutylhydantoin
δ-Bromobutylhydantoin
Formel:
C₇H₁₁BrN₂O₂ Molecular Weight: 235,081 g/mol Schmelzpunkt: 129…131 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00832189 CAS-Nummer: 28484-49-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...