4'-(Bromomethyl)-3'-chloro-[1,1'-biphenyl]-2-carbonitrile 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(4-(Bromomethyl)-3-chlorophenyl)benzonitrile, 2-Chloro-4-(2-cyanophenyl)benzyl bromide

OR8211-100MG
APOSOR8211-100MGEA 55.1 EUR
APOSOR8211-100MG
4'-(Bromomethyl)-3'-chloro-[1,1'-biphenyl]-2-carbonitrile 98%
4'-(Bromomethyl)-3'-chloro-[1,1'-biphenyl]-2-carbonitrile
Formel: C₁₄H₉BrClN
Molecular Weight: 306,585 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07777122
CAS-Nummer: 887268-24-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR