3-Phenyl-1,2,3-triazolo(1,5-a)pyridine

Lieferant: Apollo Scientific

OR8212-1G
APOSOR8212-1GEA 451 EUR
APOSOR8212-1G
3-Phenyl-1,2,3-triazolo(1,5-a)pyridine
3-Phenyl-1,2,3-triazolo(1,5-a)pyridine
Formel: C₁₂H₉N₃
Molecular Weight: 195,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08062414
CAS-Nummer: 832-81-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR