3-Pyridin-2-yl[1,2,3]triazolo[1,5-a]pyridine

Lieferant: Apollo Scientific
Achtung

OR8213-1G
APOSOR8213-1GEA 519 EUR
APOSOR8213-1G
3-Pyridin-2-yl[1,2,3]triazolo[1,5-a]pyridine
3-Pyridin-2-yl[1,2,3]triazolo[1,5-a]pyridine
Formel: C₁₁H₈N₄
Molecular Weight: 196,208 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08062393
CAS-Nummer: 947-88-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR