2-Iod-4-nitroanisol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Iodo-1-methoxy-4-nitrobenzene

OR8218-25G
APOSOR8218-25GEA 81.2 EUR
APOSOR8218-25G
2-Iod-4-nitroanisol 98%
2-Iod-4-nitroanisol
Formel: C₇H₆INO₃
Molecular Weight: 279,03 g/mol
Schmelzpunkt: 96…98 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00024328
CAS-Nummer: 5399-03-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR