6-Amino-m-kresol 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR8233-100G
APOSOR8233-100GEA
89.9
EUR
APOSOR8233-100G
6-Amino-m-kresol 98%
6-Amino-m-kresol
Formel:
C₇H₉NO Schmelzpunkt: 159…162 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007693 CAS-Nummer: 2835-98-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...