3-Hydroxy-5-nitrobenzonitril 98%

Lieferant: Apollo Scientific
Achtung

OR8249-250MG
APOSOR8249-250MGEA 33.5 EUR
APOSOR8249-250MG
3-Hydroxy-5-nitrobenzonitril 98%
3-Hydroxy-5-nitrobenzonitril
Formel: C₇H₄N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09258662
CAS-Nummer: 929000-02-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR