8-Chloro-3-methyl[1,2,4]triazolo[4,3-a]pyridine 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 8-Chloro-3-methyl-[1,2,4]triazolo[4,3-a]pyridine

OR8250-1G
APOSOR8250-1GEA 183 EUR
APOSOR8250-1G
8-Chloro-3-methyl[1,2,4]triazolo[4,3-a]pyridine 98%
8-Chloro-3-methyl[1,2,4]triazolo[4,3-a]pyridine
Formel: C₇H₆ClN₃
Molecular Weight: 167,6 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09258771
CAS-Nummer: 929000-42-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR